(R)-tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate,97% 1 * 25 g
AMBDA257926-25G
: AMBEED
Frequently Bought Together
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
: Thermo Fisher Scientific
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
: Thermo Fisher Scientific
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, liquid
: PanReac AppliChem
Diethyl ether ≥99% stabilised for HPLC
: Thermo Fisher Scientific
Cyclohexanone ≥99.0% analytical reagent
: VWR Chemicals
1,4-Dioxane, extra dry 99.8% stabilised, AcroSeal®
: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
: Thermo Fisher Scientific
Acetonitrile, anhydrous ≥99.8%
: Thermo Fisher Scientific