(2-(Methoxymethoxy)-5-methylphenyl)boronic acid,96% 1 * 0,25 g
Catalog # AMBDA742782-250MG
Supplier: AMBEED
(2-(Methoxymethoxy)-5-methylphenyl)boronic acid,96% 1 * 0,25 g
Catalog # AMBDA742782-250MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Methanol ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Hexane ≥97%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform 99.8% stabilised for spectroscopy
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
ETHYL ACETATE EXTRA PURE, DAC, N. F. 1 * 100 L
Supplier: Honeywell Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Isoamyl Alcohol according to Gerber for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem