(E)-N-(2-Chloro-6-methylphenyl)-3-ethoxyacrylamide,97% 1 * 5 g
Catalog # AMBDA297888-5G
Supplier: AMBEED
(E)-N-(2-Chloro-6-methylphenyl)-3-ethoxyacrylamide,97% 1 * 5 g
Catalog # AMBDA297888-5G
Supplier: AMBEED
Frequently Bought Together
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Toluene, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane ≥95%, TECHNICAL
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute, EMPROVE® EXPERT Ph. Eur., BP, ChP, JP, USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Pentane ≥95%, TECHNICAL
Supplier: VWR Chemicals
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl