4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine,98% 1 * 0,25 g
AMBDA173951-250MG
: AMBEED
Frequently Bought Together
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
: VWR Chemicals
Dimethyl sulphoxide ≥99.8%, SPECTRONORM® for spectroscopy
: VWR Chemicals
Acrylonitrile 99+%
: Thermo Fisher Scientific
4-Methyl-2-pentanone 99.5% for analysis
: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
Dimethyl sulphoxide, anhydrous ≥99.8%
: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
: PanReac AppliChem
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
: VWR Chemicals
Tetrahydrofuran ≥99% stabilised
: Thermo Fisher Scientific
Carbon disulphide 99.9% for HPLC
: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry 99+%, AcroSeal®
: Thermo Fisher Scientific