(2-(Methylamino)pyrimidin-5-yl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA658451-250MG
Supplier: AMBEED
(2-(Methylamino)pyrimidin-5-yl)boronic acid,98% 1 * 0,25 g
Catalog # AMBDA658451-250MG
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
N-HEPTANE 95% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
Acetone ≥99.5% ACS
Supplier: VWR Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.5% stabilised ACS
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
Methanol Reag. Ph. Eur., ACS, ISO, Burdick & Jackson™
Supplier: Honeywell Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Hexanesulphonic acid sodium salt monohydrate, Supelco®
Supplier: Merck