3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-4-amine,98+% 1 * 0,1 g
Catalog # AMBDA442760-100MG
Supplier: AMBEED
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-4-amine,98+% 1 * 0,1 g
Catalog # AMBDA442760-100MG
Supplier: AMBEED
Frequently Bought Together
Methanol ≥99.5% (by GC), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, extra pure suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
n-Heptane
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Methylpyrrolidine 98%
Supplier: Thermo Fisher Scientific
Glycerine (synthetic) 99+%, pure
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Heptane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
n-Hexane
Supplier: Roth Carl
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Diethyl ether ≥99% stabilised for HPLC
Supplier: Thermo Fisher Scientific
Glycerol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 2,5 L
Supplier: PanReac AppliChem
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
1,2-Propanediol (USP, BP, Ph. Eur., JP) pure, pharma grade 1 * 1.000 mL
Supplier: PanReac AppliChem