tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate,98% 1 * 0,1 g
AMBDA155552-100MG
: AMBEED
tert-Butyl (4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)carbamate,98% 1 * 0,1 g
AMBDA155552-100MG
: AMBEED
Frequently Bought Together
Cyclohexanone ≥99%, GPR RECTAPUR®
: VWR Chemicals
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
Toluene, GPR RECTAPUR® low in sulphur
: VWR Chemicals
Dichloromethane, anhydrous ≥99.7% stabilised
: Thermo Fisher Scientific
n-Heptane ∼ 99% (n-heptane) for HPLC
: Thermo Fisher Scientific
Chlorobenzene ≥99%
: Thermo Fisher Scientific
Pyridine ≥99.0% ACS, Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide ≥99.7% for HPLC
: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
: Honeywell Chemicals