6-BROMO-2-METHYL-3-(4,4,5,5-TETRAMETHYL- 1 * 0,25 g
Catalog # AMBDA633967-250MG
Supplier: AMBEED
Frequently Bought Together
Ethanol denatured 96% ≥96%, GPR RECTAPUR®
Supplier: VWR Chemicals
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Hexane (mixture of isomers), TECHNICAL
Supplier: VWR Chemicals
Cyclohexaan, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
(±)-2-Butaanol ≥99% analytical reagent
Supplier: VWR Chemicals
Benzoic acid methyl ester, EMPLURA®, Supelco®
Supplier: Merck
n-Hexaan, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methyleenchloride ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol Eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Aceton, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Water met 0,1% (v/v) Trifluorazijnzuur, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Tolueen, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Zoutzuur ≥36,5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals