1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole,95% 1 * 1 g
AMBDA434561-1G
: AMBEED
1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole,95% 1 * 1 g
AMBDA434561-1G
: AMBEED
Frequently Bought Together
Toluene, GPR RECTAPUR® low in sulphur
: VWR Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
: Thermo Fisher Scientific
2-Methylbutane 99+%, extra pure
: Thermo Fisher Scientific
2-Methylbutane for HPLC
: Thermo Fisher Scientific
Diethyl ether, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Chloroform ≥99.5% stabilised
: Honeywell Chemicals
n-Pentane for analysis
: Thermo Fisher Scientific
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
: Thermo Fisher Scientific
Toluene ≥99.3%, Laboratory Reagent
: Honeywell Chemicals