1-Ethyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole,95% 1 * 0,1 g
Catalog # AMBDA269240-100MG
Supplier: AMBEED
1-Ethyl-3,5-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole,95% 1 * 0,1 g
Catalog # AMBDA269240-100MG
Supplier: AMBEED
Frequently Bought Together
Benzoic acid methyl ester
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
N-Ethyldiisopropylamine ≥99.5% for peptide synthesis
Supplier: VWR Chemicals
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
1-Chloro-2,4-dinitrobenzene 99%
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Bromine 99.6% for analysis
Supplier: Thermo Fisher Scientific
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific