(R)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthalene,97% 99%ee 1 * 5 g
Catalog # AMBDA465873-5G
Supplier: AMBEED
(R)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthalene,97% 99%ee 1 * 5 g
Catalog # AMBDA465873-5G
Supplier: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran ≥99.7% unstabilised for HPLC, for UV spectrophotometry
Supplier: Thermo Fisher Scientific
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
2-Propanol USP, J.T.Baker®
Supplier: Avantor
Tetrahexylammonium bromide, Supelco®
Supplier: Merck
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
n-Hexane ≥97.0% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol (untaxed) ~96% (v/v), CHROMASOLV™ ACS for UV spectrophotometry extra fine spirit, E15 o 1, Riedel-de Haën™
Supplier: Honeywell Chemicals