tert-Butyl 3-(methylamino)pyrrolidine-1-carboxylate,97% 1 * 5 g
Catalog # AMBDA137902-5G
Supplier: AMBEED
tert-Butyl 3-(methylamino)pyrrolidine-1-carboxylate,97% 1 * 5 g
Catalog # AMBDA137902-5G
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Water, TECHNICAL, demineralised
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
Pyridine, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals