(R)-2,2-DIMETHYL-1,3-DIOXOLANE-4-CARBALD 1 * 25 g
Catalog # AMBDA852953-25G
Supplier: AMBEED
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
n-Heptaan ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methyleenchloride ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
n-Hexaan, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol Eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals