(S)-[1,1-Binaphthalene]-2,2-diyl bis(trifluoromethanesulfonate),98% 1 * 5 g
Catalog # AMBDA901332-5G
Supplier: AMBEED
(S)-[1,1-Binaphthalene]-2,2-diyl bis(trifluoromethanesulfonate),98% 1 * 5 g
Catalog # AMBDA901332-5G
Supplier: AMBEED
Frequently Bought Together
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone, EMPLURA®, Reagent Grade for synthesis, Supelco®
Supplier: Merck
Toluene, extra dry over Molecular Sieve 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…50 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific