N,N,N-TRIMETHYLTETRADECAN-1-AMINIUM BROM 1 * 25 g
Catalog # AMBDA289209-25G
Leverancier: AMBEED
Frequently Bought Together
Aceton, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Methanol ≥99.9%, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
Leverancier: Merck
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Leverancier: Merck
Ethanol absoluut ≥99.5%, EMPLURA®, Supelco®
Leverancier: Merck
2-Propanol ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Leverancier: Merck
Xyleen ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Tetrahydrofuraan, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Leverancier: Thermo Fisher Scientific