((1H-BENZO[D][1,2,3]TRIAZOL-1-YL)OXY)TRI 1 * 25 g
Catalog # AMBDA132694-25G
Supplier: AMBEED
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tolueen, EMPLURA®, extra pure, Supelco®
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Trifluorazijnzuur ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Chloorbenzeen ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, SupraSolv® for GC-MS
Supplier: Merck
Butylacetaat ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Chloroform ≥99.9% (GC, on anhydrous basis) stabilised, PESTINORM® Supra Trace
Supplier: VWR Chemicals
Ethylacetaat ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Aceton ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butaanol, watervrij ≥99.9%
Supplier: Thermo Fisher Scientific
1-Butaanol ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitril, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals