2,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole,98% 1 * 0,25 g
AMBDA513649-250MG
: AMBEED
2,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole,98% 1 * 0,25 g
AMBDA513649-250MG
: AMBEED
Frequently Bought Together
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
: PanReac AppliChem
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
: PanReac AppliChem
1,3-Dioxolane 99.5+% stabilised, pure
: Thermo Fisher Scientific
Ethanolamine ACS
: Thermo Fisher Scientific
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals
2-Nitrophenylhydrazine, moistened 30% with water 97%
: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
: Thermo Fisher Scientific
N-Methyl-2-pyrrolidone (NMP) 99.5% for analysis
: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
tert-Butanol, anhydrous ≥99.5%
: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane ≥99.5% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
: Honeywell Chemicals