2,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine,98% 1 * 0,25 g
Catalog # AMBDA883741-250MG
Supplier: AMBEED
2,3-Difluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine,98% 1 * 0,25 g
Catalog # AMBDA883741-250MG
Supplier: AMBEED
Frequently Bought Together
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Acetone, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
tert-Butanol, EMPLURA®, Supelco®
Supplier: Merck
Acetonitrile ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Hydrogen peroxide 30%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
2,2,4-Trimethylpentane 99+%, pure
Supplier: Thermo Fisher Scientific
N-HEXANE 99% CHROMAR HPLC | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND
Dichloromethane ≥99.8% stabilised, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific