(R)-1-tert-Butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate,97% 1 * 25 g
Catalog # AMBDA164851-25G
Supplier: AMBEED
(R)-1-tert-Butyl 2-methyl 5-oxopyrrolidine-1,2-dicarboxylate,97% 1 * 25 g
Catalog # AMBDA164851-25G
Supplier: AMBEED
Frequently Bought Together
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
n-Octane ≥98%
Supplier: Thermo Fisher Scientific
1,1,2,2-Tetrachloroethane 98.5%
Supplier: Thermo Fisher Scientific
Methanol 99.8% for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide, anhydrous (max. 0.005% H₂O)
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99.0%, AnalaR NORMAPUR® ACS
Supplier: VWR Chemicals
Diisopropyl ether 98+% stabilised, pure
Supplier: Thermo Fisher Scientific