(4-Bromophenyl)(phenyl)methanone,98% 1 * 100 g
AMBDA105644-100G
: AMBEED
Frequently Bought Together
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Water, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
N,N-Dimethylacetamide ≥99.5%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals