Pentafluorophenyl methacrylate,98% (stabilized with MEHQ) 1 * 25 g
Catalog # AMBDA866606-25G
Supplier: AMBEED
Pentafluorophenyl methacrylate,98% (stabilized with MEHQ) 1 * 25 g
Catalog # AMBDA866606-25G
Supplier: AMBEED
Frequently Bought Together
n-Pentane ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
DIMETHYL SULFOXIDE FOR ANALYSIS EMSURE® ACS 1 * 5 L
Supplier: Merck
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide ≥99.8% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific
1,2-Dichlorobenzene 99+% for HPLC
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Cyclohexane ≥99.7%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Chlorobenzene 99.9% for HPLC
Supplier: Thermo Fisher Scientific