N4,N4'-Diphenyl-[1,1'-biphenyl]-4,4'-diamine,98% 1 * 5 g
Catalog # AMBDA800662-5G
Supplier: AMBEED
Frequently Bought Together
2,2,4-Trimethylpentane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Methyltrichlorosilane for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
1-Propanol ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Glycerine, Hi-LR™
Supplier: HIMEDIA
Turpentine oil
Supplier: Roth Carl
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
Mixture ASTM D3230, VOLUSOL®
Supplier: VWR Chemicals
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, dried ≥99.9%, Puriss. p.a. (≤0.02% H₂O), Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals