4-((4,6-Bis(octylthio)-1,3,5-triazin-2-yl)amino)-2,6-di-tert-butylphenol,95% 1 * 5 g
Catalog # AMBDA130976-5G
Supplier: AMBEED
4-((4,6-Bis(octylthio)-1,3,5-triazin-2-yl)amino)-2,6-di-tert-butylphenol,95% 1 * 5 g
Catalog # AMBDA130976-5G
Supplier: AMBEED
Frequently Bought Together
(±)-2-Methyl-2,4-pentanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethyl acetate, EMPLURA®, Supelco®
Supplier: Merck
2,5-Di-tert-butylhydroquinone for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
4-Methyl-2-pentanone 99.5% for analysis
Supplier: Thermo Fisher Scientific
Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
BOTTLE HDPE 125ML DOSED HN03 52.5% 1.25M 1 * 84 items
Supplier: VWR Chemicals
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Dimethyl sulphoxide ≥99.5%, Reagent Grade
Supplier: Honeywell Chemicals
2-Methylpropan-1-ol GPR RECTAPUR Metal drum 25L 1 * 25 L
Supplier: VWR Chemicals