3-Methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde,95% 1 * 1 g
Catalog # AMBDA323418-1G
Supplier: AMBEED
3-Methoxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzaldehyde,95% 1 * 1 g
Catalog # AMBDA323418-1G
Supplier: AMBEED
Frequently Bought Together
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99.9% stabilised, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, TechniSolv®, pure
Supplier: VWR Chemicals
Ethanol eurodenatured ≥96%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
2-Methylbutane ≥99%
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butylboronic acid, Supelco®
Supplier: Merck
Diethylene glycol monobutyl ether ≥98%, TECHNICAL
Supplier: VWR Chemicals
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific