(2S,3aS,7aS)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid,98% 1 * 5 g
Catalog # AMBDA165139-5G
Supplier: AMBEED
(2S,3aS,7aS)-1-(((9H-Fluoren-9-yl)methoxy)carbonyl)octahydro-1H-indole-2-carboxylic acid,98% 1 * 5 g
Catalog # AMBDA165139-5G
Supplier: AMBEED
Frequently Bought Together
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Water with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
Diethylene glycol monobutyl ether, Sigma-Aldrich®
Supplier: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Diethyl ether, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
Ethanol absolute (untaxed) ≥99.8% (by GC), Puriss., Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclopentane, extra dry 95+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals