Sodium 3-(4-hydroxy-3-methoxyphenyl)acrylate,99+% 1 * 0,25 g
Catalog # AMBDA203361-250MG
Supplier: AMBEED
Sodium 3-(4-hydroxy-3-methoxyphenyl)acrylate,99+% 1 * 0,25 g
Catalog # AMBDA203361-250MG
Supplier: AMBEED
Frequently Bought Together
2,3-Dimercapto-1-propanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Anisole 99%, Reagent Grade
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
Supplier: Thermo Fisher Scientific
Toluene ≥99.5% ACS
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
tert-Butyl methyl ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, extra pure
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane 99.5% for analysis
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific