2-Ethoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol,97% GC (contain isomer) 1 * 25 g
Catalog # AMBDA226554-25G
Supplier: AMBEED
2-Ethoxy-4-(4-methyl-1,3-dioxolan-2-yl)phenol,97% GC (contain isomer) 1 * 25 g
Catalog # AMBDA226554-25G
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate anhydrous, 99.8 % 1 * 2 L
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
(±)-Tetrahydrofurfuryl alcohol, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.9% (by GC) Reag. Ph. Eur., ACS, ISO for analysis
Supplier: PanReac AppliChem
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
1-Pentanol ≥98%
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
n-Hexane
Supplier: Roth Carl
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor