N1,N1'-(Ethane-1,2-diyl)bis(N1,N2,N2-trimethylethane-1,2-diamine),97% 1 * 5 g
Catalog # AMBDA407765-5G
Supplier: AMBEED
N1,N1'-(Ethane-1,2-diyl)bis(N1,N2,N2-trimethylethane-1,2-diamine),97% 1 * 5 g
Catalog # AMBDA407765-5G
Supplier: AMBEED
Frequently Bought Together
Hexane anhydrous, 95 % 1 * 250 mL
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Chloroform 99.0-99.4% (by GC) stabilised, Puriss. p.a. ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile 99.9%, ECD tested for residue analysis
Supplier: Thermo Fisher Scientific
Petroleum spirit, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol 99+% ACS
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND