3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride,99+% 1 * 100 g
Catalog # AMBDA241911-100G
Supplier: AMBEED
3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride,99+% 1 * 100 g
Catalog # AMBDA241911-100G
Supplier: AMBEED
Frequently Bought Together
Dipentyl ether stabilised, Sigma-Aldrich®
Supplier: Merck
(1S)-(-)-α-Pinene for synthesis, Sigma-Aldrich®
Supplier: Merck
Benzoic acid methyl ester, EMPLURA®, Supelco®
Supplier: Merck
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
1-Naphthol for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
Diiodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
FAM Benzine DIN 51635, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Anisole, extra dry 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Trichloroacetic Acid (BP, Ph. Eur.) pure, pharma grade 1 * 5 kg
Supplier: PanReac AppliChem