6-Hydroxy-2-naphthaldehyde,98% 1 * 10 g
Catalog # AMBDA229246-10G
Supplier: AMBEED
Frequently Bought Together
Chloroform, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
n-Hexane ≥99.0% (by GC), LiChrosolv®, hypergrade for LC-MS
Supplier: Merck
Acetone ≥99.8%, SupraSolv® for GC-MS, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
n-Heptane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Pentane, EMSURE® for analysis, Supelco®
Supplier: Merck
Cyclohexane ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
2-Propanol, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals