(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,25 g
Catalog # AMBDA1719179-250MG
Supplier: AMBEED
(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,25 g
Catalog # AMBDA1719179-250MG
Supplier: AMBEED
Frequently Bought Together
Methyl acetate anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
Tetraheptylammonium bromide, Supelco®
Supplier: Merck
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Methanol ≥99.9% ACS, Spectrophotometric Grade for spectrophotometry, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol denatured ≥96%, meets analytical specification of Ph.Eur. (denaturated with 1%iPr-OH,1% MEK,0.001% Bitrex®)
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
N,N-Dimethylacetamide 99%, pure
Supplier: Thermo Fisher Scientific