(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,1 g
Catalog # AMBDA1665968-100MG
Supplier: AMBEED
(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,1 g
Catalog # AMBDA1665968-100MG
Supplier: AMBEED
Frequently Bought Together
Dimethyl sulphoxide, EMPLURA®, Supelco®
Supplier: Merck
n-Hexane ≥99.0%, PESTINORM® for capillary GC analysis
Supplier: VWR Chemicals
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Iodomethane stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Ethyl acetate, EMPLURA®, Supelco®
Supplier: Merck
n-Heptane
Supplier: Merck
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Xylene (mixture of isomers) EMPARTA® ACS analytical reagent
Supplier: Merck
Diethyl ether, anhydrous (max. 0.005% H₂O) ≥99.8% stabilised
Supplier: VWR Chemicals
Carbon disulphide 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Chloroform 99.0-99.4% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,3-Dioxolane 99.5+% stabilised, pure
Supplier: Thermo Fisher Scientific
1-Octanol ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Ethyl Acetate (BP, Ph. Eur.) pharma grade 1 * 200 L
Supplier: PanReac AppliChem