(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,25 g
Catalog # AMBDA1672154-250MG
Supplier: AMBEED
(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,25 g
Catalog # AMBDA1672154-250MG
Supplier: AMBEED
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Phenol liquified ≥90% for synthesis
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
Pyridine, extra dry over Molecular Sieve 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chloroform ≥99.8% stabilised ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
tert-Butanol, anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Pyridine ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
1,2-Dimethoxyethane, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethanolamine ACS
Supplier: Thermo Fisher Scientific