(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,1 g
Catalog # AMBDA1670279-100MG
Supplier: AMBEED
(R)-4-(tert-Butyl)-2-(2-(diphenylphosphanyl)phenyl)-5,5-diphenyl-4,5-dihydrooxazole,98% 1 * 0,1 g
Catalog # AMBDA1670279-100MG
Supplier: AMBEED
Frequently Bought Together
Acetonitrile, Prepsolv® for preparative chromatography
Supplier: Merck
Ethyl acetate, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
ACETONITRILE EMPLURA® 1 * 4 L
Supplier: Merck
Methanol ≥99.8%, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
Water, extra pure, Deionized
Supplier: Thermo Fisher Scientific
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.8% ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Ethyl acetate, AR®, Laboratory Reagent, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Acetonitrile, extra dry 99.9+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific
Methanol 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Cyclohexane for pesticide residue analysis
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran ≥99.9% inhibitor-free, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals