2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-yl methanesulfonate,95% 1 * 0,25 g
Catalog # AMBDA1523589-250MG
Supplier: AMBEED
2,2-Dimethyl-4-oxo-3,8,11-trioxa-5-azatridecan-13-yl methanesulfonate,95% 1 * 0,25 g
Catalog # AMBDA1523589-250MG
Supplier: AMBEED
Frequently Bought Together
tert-Butyl methyl ether ≥99.0% (by GC), EMPLURA®, Supelco®
Supplier: Merck
N-Hydroxysuccinimide for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene - Mixture of isomers technical grade 1 * 5 L
Supplier: PanReac AppliChem
Tetrahydrofuran stabilised ACS, Burdick & Jackson™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Toluene, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
1,4-Dioxane 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
2-Propanol ≥99.8% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Polyvinyl alcohol, M.W. ∼ 13,000 98%, hydrolysed
Supplier: Thermo Fisher Scientific
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem