(S)-2-(5-(Bis(4-chlorophenyl)methyl)-3-methylthiophene-2-carboxamido)-5-guanidinopentanoic acid,95% 1 * 0,25 g
Catalog # AMBDA1532786-25MG
Supplier: AMBEED
(S)-2-(5-(Bis(4-chlorophenyl)methyl)-3-methylthiophene-2-carboxamido)-5-guanidinopentanoic acid,95% 1 * 0,25 g
Catalog # AMBDA1532786-25MG
Supplier: AMBEED
Frequently Bought Together
Water with 0,1% (v/v) formic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals