1-((3S,4R)-4-(2,6-Difluoro-4-methoxyphenyl)-2-oxopyrrolidin-3-yl)-3-phenylurea,98+% 1 * 0,25 g
Catalog # AMBDA1538642-250MG
Supplier: AMBEED
1-((3S,4R)-4-(2,6-Difluoro-4-methoxyphenyl)-2-oxopyrrolidin-3-yl)-3-phenylurea,98+% 1 * 0,25 g
Catalog # AMBDA1538642-250MG
Supplier: AMBEED
Frequently Bought Together
n-Hexane ≥98%, SupraSolv® for GC-MS
Supplier: Merck
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.5% (by GC), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, extra pure suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethylene glycol for synthesis
Supplier: Merck
Dimethyl sulphoxide ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Triethylamine ≥99.6%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific