1,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole,95% 1 * 0,1 g
Catalog # AMBDA1522786-100MG
Supplier: AMBEED
1,4-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole,95% 1 * 0,1 g
Catalog # AMBDA1522786-100MG
Supplier: AMBEED
Frequently Bought Together
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, CHROMASOLV™, gradient grade for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether stabilised, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals