1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate,97% 1 * 0,25 g
AMBDA1496205-250MG
: AMBEED
1-(tert-Butyl) 4-(1,3-dioxoisoindolin-2-yl) piperidine-1,4-dicarboxylate,97% 1 * 0,25 g
AMBDA1496205-250MG
: AMBEED
Frequently Bought Together
Ethanol absolute ≥99.5%, TechniSolv®, pure
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Cyclohexanone ≥99%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone, J.T.Baker®
: Avantor
Glycerine, anhydrous ≥99.5% (by GC), Puriss. p.a. ACS, distilled, Riedel-de Haën™
: Honeywell Chemicals