(2R,3R)-3-((R)-1-((tert-Butyldimethylsilyl)oxy)ethyl)-4-oxoazetidin-2-yl acetate,99+% 1 * 5 g
AMBDA1473715-5G
: AMBEED
(2R,3R)-3-((R)-1-((tert-Butyldimethylsilyl)oxy)ethyl)-4-oxoazetidin-2-yl acetate,99+% 1 * 5 g
AMBDA1473715-5G
: AMBEED
Frequently Bought Together
Acetonitrile for DNA synthesis
: Merck
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Cesium chloride 99.99% (trace metals basis), Optical Grade for spectroscopy
: Thermo Fisher Scientific
1,4-Dioxane 99+% ACS
: Thermo Fisher Scientific
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
4-(Methylamino)phenol sulphate 99%
: Thermo Fisher Scientific
Methanol dry (max. 0.005% water) , ACS, ISO 1 * 1 L
: PanReac AppliChem
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
Ethanol (untaxed) in water ≥70% (v/v) Ph. Helv., Riedel-de Haën™
: Honeywell Chemicals
Cyclohexane ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals