4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride,Indicator(pH 1.0-3.1) 1 * 25 g
AMBDA1458376-25G
: AMBEED
4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride,Indicator(pH 1.0-3.1) 1 * 25 g
AMBDA1458376-25G
: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
: Merck
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Tetrachloroethylene ≥98%, TECHNICAL
: VWR Chemicals
Dimethyl sulphoxide ≥99%
: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
: Honeywell Chemicals
N,N-Dimethylformamide 99+%, extra pure
: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
: Thermo Fisher Scientific
ETHYLENE GLYCOL ≥98 TECHNICAL 1 * 5 L
: Roth Carl
Diisopropyl ether 98+% stabilised, pure
: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
: Thermo Fisher Scientific