(2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol,98% 1 * 0,1 g
Catalog # AMBDA410468-100MG
Supplier: AMBEED
(2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol,98% 1 * 0,1 g
Catalog # AMBDA410468-100MG
Supplier: AMBEED
Frequently Bought Together
Diisopropyl ether 99+% stabilised, extra pure
Supplier: Thermo Fisher Scientific
Acetone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
Methanol ≥99.9%, CHROMASOLV™ for capillary GC analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Glycerine ≥83.5-88.5% (w/w) in aqueous solution, pure
Supplier: Thermo Fisher Scientific
Methyl ethyl ketone, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals
Dichloromethane
Supplier: Roth Carl
4-(Methylamino)phenol sulphate 99%
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
1-Pentanol 99%, pure
Supplier: Thermo Fisher Scientific
Toluene, anhydrous ≥99.8%, over molecular sieves
Supplier: Thermo Fisher Scientific