2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one,97% 1 * 1 g
Catalog # AMBDA121511-1G
Supplier: AMBEED
2-Amino-8-bromo-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1H-purin-6(9H)-one,97% 1 * 1 g
Catalog # AMBDA121511-1G
Supplier: AMBEED
Frequently Bought Together
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Ethanol absolute ≥99.8% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Chloroform, extra dry over Molecular Sieve 99.9% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
2-Methylbutane 99+%, extra pure
Supplier: Thermo Fisher Scientific
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Toluene ≥99.3%, Laboratory Reagent
Supplier: Honeywell Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
1,4-Dioxane, extra dry over Molecular Sieve 99,5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.9%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Isoamyl alcohol ≥98.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals