2-Bromo-4-(perfluoropropan-2-yl)-6-(trifluoromethyl)aniline,98% 1 * 0,1 g
Catalog # AMBDA1273251-100MG
Supplier: AMBEED
2-Bromo-4-(perfluoropropan-2-yl)-6-(trifluoromethyl)aniline,98% 1 * 0,1 g
Catalog # AMBDA1273251-100MG
Supplier: AMBEED
Frequently Bought Together
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
Benzyl alcohol ≥99.0% analytical reagent
Supplier: VWR Chemicals
1,3-Dimethyl-2-imidazolidinone ≥99.5%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Diethyl ether BHT 1ppm inhibitor, anhydrous, 99.7 % 1 * 1 L
Supplier: Merck
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
Ethylenediamine 99%
Supplier: Thermo Fisher Scientific
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.5% ACS
Supplier: Thermo Fisher Scientific
Water ACS, Deionized
Supplier: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Supplier: Merck
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific