(S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-6-fluoro-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,97% 1 * 0,1 g
AMBDA1258571-100MG
: AMBEED
(S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-6-fluoro-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid,97% 1 * 0,1 g
AMBDA1258571-100MG
: AMBEED
Frequently Bought Together
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Glycerine (glycerol) USP, Ph. Eur., BP
: VWR Chemicals
Petroleum spirit, 50…70 °C ≥90%, AnalaR NORMAPUR®
: VWR Chemicals
Tetrachloroethylene 99%, extra pure
: Thermo Fisher Scientific
Iodomethane ≥99% stabilised
: Thermo Fisher Scientific
N,N-Dimethylacetamide 99.8+% for HPLC
: Thermo Fisher Scientific
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
: Honeywell Chemicals