(11bR)-N,N-Bis((1S)-1-phenylethyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine,98% 99%ee 1 * 0,1 g
Catalog # AMBDA1188217-100MG
Supplier: AMBEED
(11bR)-N,N-Bis((1S)-1-phenylethyl)dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine,98% 99%ee 1 * 0,1 g
Catalog # AMBDA1188217-100MG
Supplier: AMBEED
Frequently Bought Together
n-Heptane, EMPLURA®, Supelco®
Supplier: Merck
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
Methanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Pyridine dry (max. 0.01% water) , ACS 1 * 1.000 mL
Supplier: PanReac AppliChem
Cyclohexane 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Ethylene glycol ≥99% (by GC), pure
Supplier: PanReac AppliChem
Isoamyl alcohol mixture of isomers ≥99%
Supplier: Thermo Fisher Scientific
Acetonitrile 99.9% ACS, gradient grade, meets analytical specification of Ph.Eur. for HPLC
Supplier: Thermo Fisher Scientific
Petroleum spirit, 40…60 °C, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Toluene 99.8+% for analysis
Supplier: Thermo Fisher Scientific