2-(2-Bromophenoxy)-N,N-dimethylacetamide,95% 1 * 0,25 g
Catalog # AMBDA1061323-250MG
Supplier: AMBEED
2-(2-Bromophenoxy)-N,N-dimethylacetamide,95% 1 * 0,25 g
Catalog # AMBDA1061323-250MG
Supplier: AMBEED
Frequently Bought Together
1,3-Propanediol ≥99.7%, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
N,N-Dimethylformamide ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Diethyl ether, EMPARTA® ACS for analysis, Supelco®
Supplier: Merck
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol ≥96% (v/v)
Supplier: KLINIPATH
2-Propanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Diethylene glycol for synthesis
Supplier: Merck
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Pyridine ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Diethyl ether ≥99.5% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
tert-Butyl methyl ether, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific