N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-phenyl-9H-fluoren-2-amine,95% 1 * 0,25 g
Catalog # AMBDA989538-250MG
Supplier: AMBEED
N-(9,9-Dimethyl-9H-fluoren-2-yl)-9,9-dimethyl-N-phenyl-9H-fluoren-2-amine,95% 1 * 0,25 g
Catalog # AMBDA989538-250MG
Supplier: AMBEED
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, PFAS grade for LC-MS
Supplier: VWR Chemicals
Benzoic acid methyl ester
Supplier: Merck
Cyclohexanone, EMPLURA®, extra pure, Supelco®
Supplier: Merck
Toluene, extra dry 99.85%, AcroSeal®
Supplier: Thermo Fisher Scientific
p-Xylene 99% for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (GC, on anhydrous basis) stabilised for peptide synthesis
Supplier: VWR Chemicals
(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Xylene - Mixture of isomeres (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
n-Hexane 95+%, extra pure
Supplier: Thermo Fisher Scientific
o-Xylene ≥99.0% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
n-Heptane ∼ 99% (n-heptane) for HPLC
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide (DMSO),?99,9 %, Ph. Eur., USP 1 * 500 mL
Supplier: Roth Carl
4,4'-Bis(dimethylamino)diphenylmethane 98%
Supplier: Thermo Fisher Scientific