2,2,3,3,9,9,10,10-Octamethyl-4,8-dioxa-3,9-disilaundecan-6-ol,90% 1 * 25 g
AMBDA834475-25G
: AMBEED
Frequently Bought Together
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals