2-Ethoxy-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine,97% 1 * 0,1 g
AMBDA676431-100MG
: AMBEED
2-Ethoxy-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine,97% 1 * 0,1 g
AMBDA676431-100MG
: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals